Check Order Status
Sodium Hydroxide, Certified AR for Analysis, 32% Solution, for Nitrogen Determination, Fisher Chemical™
Element Search
Structure Search
Sodium Hydroxide, Certified AR for Analysis, 32% Solution, for Nitrogen Determination, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FGCS4955PB17
also known as FGCS/4955/PB17
Price (MYR)
478.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
478.00
EA
Add to cart
Specifications
pH
14
Color
Colorless
Boiling Point
120°C
CAS
1310-73-2
Molecular Formula
HNaO
View more