Sodium Hydroxide, Certified AR for Analysis, Pearl, Fisher Chemical
Sodium Hydroxide, Certified AR for Analysis, Pearl, Fisher Chemical
Actual product may vary
Sodium Hydroxide, Certified AR for Analysis, Pearl, Fisher Chemical
Sodium Hydroxide, Certified AR for Analysis, Pearl, Fisher Chemical
Sodium Hydroxide, Certified AR for Analysis, Pearl, Fisher Chemical
Have Questions?
Catalog NumberQuantity
FGCS484560
also known as FGCS/4845/60
1 kg
Catalog number FGCS484560
also known as FGCS/4845/60
Price (MYR)
307.00
EA
Add to cart
Quantity:
1 kg
Price (MYR)
307.00
EA
Add to cart
1kg, Plastic powder jar
Specifications
pH14
ColorWhite
Boiling Point1390°C
CAS1310-73-2
Melting Point318°C
Molecular FormulaHNaO
MDL Number3548
Synonymsodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity1 kg
SMILES[OH-].[Na+]
IUPAC Namesodium;hydroxide
Molecular Weight (g/mol)39.997
PubChem CID14798
ChEBICHEBI:32145
Formula Weight40g/mol
PackagingPlastic powder jar
Vapor Pressure1mbar 700
Physical FormSolid
Unit SizeEA