Check Order Status
Sodium Hydroxide, Technical, Pearl, Fisher Chemical™
Element Search
Structure Search
Sodium Hydroxide, Technical, Pearl, Fisher Chemical™
Have Questions?
Change view
Quantity:
5 kg
1 kg
Packaging:
Plastic powder jar
Catalog number
FGCS480065
also known as FGCS/4800/65
Price (MYR)
479.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
479.00
EA
Add to cart
Specifications
pH
14
Color
White
Boiling Point
1390°C
CAS
1310-73-2
Melting Point
318°C
View more