Check Order Status
Silica Gel 60 Angstrom Pore Size, 35-70 Micron, for Column Chromatography, Fisher Chemical™
Actual product may vary
Silica Gel 60 Angstrom Pore Size, 35-70 Micron, for Column Chromatography, Fisher Chemical™
Have Questions?
Change view
Catalog Number
Packaging
Quantity
FGCS069365
also known as FGCS/0693/65
Plastic Keg
5 kg
FGCS069360
also known as FGCS/0693/60
Plastic Powder Jar
1 kg
2 Options
Catalog number
FGCS069365
also known as FGCS/0693/65
Price (MYR)
2,557.00
EA
Add to cart
Packaging:
Plastic Keg
Quantity:
5 kg
Price (MYR)
2,557.00
EA
Add to cart
Silica Gel 60 Angstrom Pore Size, 35-70 Micron, for Column Chromatography
Specifications
Packaging
Plastic Keg
Quantity
5 kg