Check Order Status
Semicarbazide Hydrochloride, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Semicarbazide Hydrochloride, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCS056048
also known as FGCS/0560/48
100 g
Plastic powder jar
Catalog number
FGCS056048
also known as FGCS/0560/48
Price (MYR)
-
Quantity:
100 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS
563-41-7
Melting Point
170°C
Color
White
pH
1.5
Packaging
Plastic powder jar
View more