Check Order Status
Salicylic Acid, 99.9+%, Certified AR for Analysis, Fisher Chemical
Element Search
Structure Search
Salicylic Acid, 99.9+%, Certified AR for Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCS028253
also known as FGCS/0282/53
500 g
Plastic powder jar
Catalog number
FGCS028253
also known as FGCS/0282/53
Price (MYR)
553.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
553.00
BOT
Add to cart
Specifications
CAS
69-72-7
Melting Point
158°C
Color
White
pH
3.6
Boiling Point
211°C
View more