Check Order Status
Safranine, Pure, C.I.50240, Fisher Chemical
Element Search
Structure Search
Safranine, Pure, C.I.50240, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCS012046
also known as FGCS/0120/46
25 g
Glass Jar
Catalog number
FGCS012046
also known as FGCS/0120/46
Price (MYR)
443.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass Jar
Request bulk or custom format
Price (MYR)
443.00
EA
Add to cart
Specifications
CAS
477-73-6
Color
Brown
Packaging
Glass Jar
Molecular Formula
C20H19ClN4
MDL Number
MFCD00011759
View more