Check Order Status
Saccharin, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Saccharin, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCS004048
also known as FGCS/0040/48
100 g
Plastic powder jar
Catalog number
FGCS004048
also known as FGCS/0040/48
Price (MYR)
1,802.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
1,802.00
EA
Add to cart
Specifications
CAS
81-07-2
Melting Point
226°C
Color
White
pH
2
Packaging
Plastic powder jar
View more