Check Order Status
Resazurin Tablets, Technical, Fisher Chemical™
Element Search
Structure Search
Resazurin Tablets, Technical, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCR004079
also known as FGCR/0040/79
100 Tablets
Catalog number
FGCR004079
also known as FGCR/0040/79
Price (MYR)
-
Quantity:
100 Tablets
Request bulk or custom format
Specifications
CAS
550-82-3
Molecular Formula
C12H7NO4
MDL Number
66739
InChI Key
PLXBWHJQWKZRKG-UHFFFAOYSA-N
SMILES
C1=CC2=C(C=C1O)OC3=CC(=O)C=CC3=[N+]2[O-]
View more