Check Order Status
Potassium Persulfate, for Electrophoresis, Fisher Chemical
Element Search
Structure Search
Potassium Persulfate, for Electrophoresis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCPP56045
also known as FGCP/P560/45
10 g
Glass Jar
Catalog number
FGCPP56045
also known as FGCP/P560/45
Price (MYR)
-
Quantity:
10 g
Packaging:
Glass Jar
Request bulk or custom format
Specifications
Melting Point
100°C
CAS
7727-21-1
Color
White
pH
4 to 5
Physical Form
Solid
View more