Check Order Status
Pyrogallol, Certified AR for Analysis, Fisher Chemical
Element Search
Structure Search
Pyrogallol, Certified AR for Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCP812248
also known as FGCP/8122/48
100 g
Glass Jar
Catalog number
FGCP812248
also known as FGCP/8122/48
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass Jar
Request bulk or custom format
Specifications
CAS
87-66-1
Melting Point
131°C
Color
White
pH
5.8
Boiling Point
309°C
View more