Check Order Status
Isopropanol 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Element Search
Structure Search
Isopropanol 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCP750115
also known as FGCP/7501/15
1 L
Catalog number
FGCP750115
also known as FGCP/7501/15
Price (MYR)
-
Quantity:
1 L
Request bulk or custom format
Specifications
pH
7
Melting Point
-89.5°C
CAS
67-63-0
Color
Colorless
Boiling Point
81°C
View more