Check Order Status
Isopropanol, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Isopropanol, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
25 L
500 mL
Packaging:
View all
Plastic Container
Catalog number
FGCP7490PC25
also known as FGCP/7490/PC25
Price (MYR)
1,784.00
EA
Add to cart
Quantity:
25 L
Packaging:
Plastic Container
Request bulk or custom format
Price (MYR)
1,784.00
EA
Add to cart
Specifications
pH
7
Melting Point
-89.5°C
CAS
67-63-0
Color
Colorless
Boiling Point
81°C
View more
Isopropanol, Extra Pure, SLR