Check Order Status
Phthalic Anhydride, 99+%, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Phthalic Anhydride, 99+%, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCP336053
also known as FGCP/3360/53
500 g
Catalog number
FGCP336053
also known as FGCP/3360/53
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Packaging
Plastic powder jar
CAS
85-44-9
Vapor Pressure
0.01mbar at 20°C
Physical Form
Solid
Molecular Formula
C8H4O3
View more