Check Order Status
Phenol Liquified 80% (w/w) In Water, Certified AR, for Analysis, Fisher Chemical™
Actual product may vary
Phenol Liquified 80% (w/w) In Water, Certified AR, for Analysis, Fisher Chemical™
C6H6O, CAS Number-108-95-2, benzenol, carbolic acid, hydroxybenzene, monophenol, oxybenzene, phenic acid, phenyl hydrate, phenyl hydroxide, phenylic acid, phenylic alcohol, 2.5L, CHEBI:15882, Red, 94.0414g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 40.5 deg.C, 2143, 94.113, Plastic Coated Glass bottle
Have Questions?
Catalog Number
Quantity
Packaging
FGCP2316PB17
also known as FGCP/2316/PB17
2.5 L
Plastic Coated Glass bottle
Catalog number
FGCP2316PB17
also known as FGCP/2316/PB17
Price (MYR)
981.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Price (MYR)
981.00
EA
Add to cart
C6H6O, CAS Number-108-95-2, benzenol, carbolic acid, hydroxybenzene, monophenol, oxybenzene, phenic acid, phenyl hydrate, phenyl hydroxide, phenylic acid, phenylic alcohol, 2.5L, CHEBI:15882, Red, 94.0414g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 40.5 deg.C, 2143, 94.113, Plastic Coated Glass bottle
Specifications
CAS
108-95-2
Melting Point
40.5°C
Color
Clear colorless mobile liquid
Packaging
Plastic Coated Glass bottle
Molecular Formula
C6H6O
MDL Number
MFCD00002143
Quantity
2.5 L
Synonym
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
InChI Key
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1
IUPAC Name
phenol
Molecular Weight (g/mol)
94.11
PubChem CID
996
Vapor Density
3.2
ChEBI
CHEBI:15882
Formula Weight
94.0414g/mol
Vapor Pressure
1mmHg 40
Physical Form
Liquid