Check Order Status
Phenol Liquified 80% (w/w) In Water, Pure, Fisher Chemical
Element Search
Structure Search
Phenol Liquified 80% (w/w) In Water, Pure, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCP2315PB17
also known as FGCP/2315/PB17
2.5 L
Plastic Coated Glass bottle
Catalog number
FGCP2315PB17
also known as FGCP/2315/PB17
Price (MYR)
965.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Price (MYR)
965.00
EA
Add to cart
Specifications
CAS
108-95-2
Melting Point
40.5°C
Color
Red
Packaging
Plastic Coated Glass bottle
Molecular Formula
C6H6O
View more