Check Order Status
Petroleum Ether 40-60°C, for Residue Analysis, Distol™, Fisher Chemical
Element Search
Structure Search
Petroleum Ether 40-60°C, for Residue Analysis, Distol™, Fisher Chemical
Have Questions?
Catalog number
FGCP209215
also known as FGCP/2092/15
Price (MYR)
482.00
Price (MYR)
482.00
EA
Add to cart
Specifications
CAS
64742-49-0
Color
Colorless
Boiling Point
40°C
Quantity
1 L
Packaging
Amber Glass Bottle
View more