Check Order Status
Petroleum Ether, Pure, (Petroleum Spirit with <15% N-Hexane), Fisher Chemical™
Element Search
Structure Search
Petroleum Ether, Pure, (Petroleum Spirit with <15% N-Hexane), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCP136017
also known as FGCP/1360/17
2.5 L
Amber Glass Bottle
Catalog number
FGCP136017
also known as FGCP/1360/17
Price (MYR)
-
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
CAS
8032-32-4
Color
Colorless
Boiling Point
38°C
Quantity
2.5 L
Packaging
Amber Glass Bottle
View more
Petroleum Ether, Pure, (Petroleum Spirit with <15% N-Hexane)