Check Order Status
Perchloric Acid, 60%, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Perchloric Acid, 60%, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Catalog number
FGCP1200PB17
also known as FGCP/1200/PB17
Price (MYR)
1,673.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,673.00
EA
Add to cart
Specifications
pH
0.1
Color
Colorless
CAS
7601-90-3
Melting Point
-18°C
Boiling Point
203°C
View more