Check Order Status
Iso-Pentane, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Iso-Pentane, Certified AR for Analysis, Fisher Chemical™
C5H12, CAS Number-78-78-4, butane, 2-methyl, 1,1-dimethylpropane, dimethylethylmethane, butanes, 1,1,2-trimethylethane, iso-pentane, iso-c5h12, isoamylhydride, ethyldimethylmethane, isopentane, 500mL, 30 deg.C, CHEBI:30362, Colorless, 72.15g/mol, QWTDNUCVQCZILF-UHFFFAOYSA-N, 2-methylbutane
Have Questions?
Catalog Number
Quantity
Packaging
FGCP103108
also known as FGCP/1031/08
500 mL
Amber glass bottle
Catalog number
FGCP103108
also known as FGCP/1031/08
Price (MYR)
320.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
320.00
EA
Add to cart
Specifications
Chemical Name or Material
Iso-Pentane
CAS
78-78-4
Melting Point
-160°C
Color
Colorless
Boiling Point
30°C
View more
C5H12, CAS Number-78-78-4, butane, 2-methyl, 1,1-dimethylpropane, dimethylethylmethane, butanes, 1,1,2-trimethylethane, iso-pentane, iso-c5h12, isoamylhydride, ethyldimethylmethane, isopentane, 500mL, 30 deg.C, CHEBI:30362, Colorless, 72.15g/mol, QWTDNUCVQCZILF-UHFFFAOYSA-N, 2-methylbutane