Check Order Status
N-Pentane, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
N-Pentane, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
5 L
500 mL
2.5 L
Packaging:
View all
Metal can
Catalog number
FGCP102021
also known as FGCP/1020/21
Price (MYR)
865.00
EA
Add to cart
Quantity:
5 L
Packaging:
Metal can
Request bulk or custom format
Price (MYR)
865.00
EA
Add to cart
Specifications
Packaging
Metal can
CAS
109-66-0
Vapor Pressure
573mbar at 20°C
Viscosity
0.25 mPaS at 20°C
Vapor Density
2.5
View more
N-Pentane, Extra Pure, SLR