Check Order Status
Pectin, Pure, Citrus-121 Grade, Slow Setting 121, Fisher Chemical
Element Search
Structure Search
Pectin, Pure, Citrus-121 Grade, Slow Setting 121, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCP092048
also known as FGCP/0920/48
100 g
Plastic powder jar
Catalog number
FGCP092048
also known as FGCP/0920/48
Price (MYR)
-
Quantity:
100 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
Color
White
pH
2.8 to 3.4
CAS
9000-69-5
Physical Form
Powder Solid
MDL Number
81838
View more