Check Order Status
Oxalic Acid Anhydrous, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Oxalic Acid Anhydrous, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCO058053
also known as FGCO/0580/53
500 g
Plastic powder jar
Catalog number
FGCO058053
also known as FGCO/0580/53
Price (MYR)
-
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
pH
1.3
CAS
144-62-7
Melting Point
189°C
Color
White
Molecular Formula
C2H2O4
View more