Orthophosphoric Acid, 85%, HPLC for Electrochemical Detection, Certified, Fisher Chemical™
Orthophosphoric Acid, 85%, HPLC for Electrochemical Detection, Certified, Fisher Chemical™
Actual product may vary
Orthophosphoric Acid, 85%, HPLC for Electrochemical Detection, Certified, Fisher Chemical™
Orthophosphoric Acid, 85%, HPLC for Electrochemical Detection, Certified, Fisher Chemical™
Orthophosphoric Acid, 85%, HPLC for Electrochemical Detection, Certified, Fisher Chemical™
H3O4P, CAS Number-7664-38-2, acide phosphorique, evits, acido fosforico, white, acidum phosphoricum, sonac, o-phosphoric acid, wc-reiniger, orthophosphoric acid, phosphorsaeure, 500mL, 158 deg.C, CHEBI:26078, Undesignated, NBIIXXVUZAFLBC-UHFFFAOYSA-N, 21 deg.C, 11340, 97.994 | CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FGCO0515PB08
also known as FGCO/0515/PB08
500 mLPlastic Coated Glass bottle
Catalog number FGCO0515PB08
also known as FGCO/0515/PB08
Price (MYR)
517.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Price (MYR)
517.00
BOT
Add to cart
H3O4P, CAS Number-7664-38-2, acide phosphorique, evits, acido fosforico, white, acidum phosphoricum, sonac, o-phosphoric acid, wc-reiniger, orthophosphoric acid, phosphorsaeure, 500mL, 158 deg.C, CHEBI:26078, Undesignated, NBIIXXVUZAFLBC-UHFFFAOYSA-N, 21 deg.C, 11340, 97.994
Specifications
Boiling Point158°C
Melting Point21°C
CAS7664-38-2
pH2
Physical FormLiquid
Quantity500 mL
Molecular FormulaH3O4P
MDL Number11340
Synonymorthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
InChI KeyNBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILESOP(=O)(O)O
IUPAC Namephosphoric acid
Molecular Weight (g/mol)97.994
PubChem CID1004
ChEBICHEBI:26078
PackagingPlastic Coated Glass bottle
Vapor Pressure2 hPa at 20°C
DensityVapor Density: 3.4
Unit SizeBOT