Check Order Status
Orcein Staining, Pure, Fisher Chemical
Element Search
Structure Search
Orcein Staining, Pure, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCO030044
also known as FGCO/0300/44
5 g
Catalog number
FGCO030044
also known as FGCO/0300/44
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS
1400-62-0
Molecular Formula
C28H24N2O7
MDL Number
62310
InChI Key
VPEASJIRGSVXBF-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(=C1NC2=C(C3=C4C(=C(C(=O)C=C4OC3=CC2=O)NC5=C(C=C(C=C5C)O)O)C)C)O)O
View more