Nitric Acid 68% d=1.42, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
Nitric Acid 68% d=1.42, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
Nitric Acid 68% d=1.42, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
Nitric Acid 68% d=1.42, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
Nitric Acid 68% d=1.42, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
HNO3, CAS Number-7697-37-2, hydrogen nitrate, nital, aqua fortis, azotic acid, acide nitrique, acidum nitricum, rfna, salpetersaeure, nitrous fumes, nitryl hydroxide, 2.5L, CHEBI:48107, Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, HDPE plastic bottle | CAS: 7697-37-2 | HNO3 | 63.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Catalog number FGCN2272PB17
also known as FGCN/2272/PB17
Price (MYR)
409.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
409.00
EA
Add to cart
Specifications
Chemical Name or MaterialNitric Acid
pH1
ColorYellow
CAS7697-37-2
Melting Point-41°C
View more
HNO3, CAS Number-7697-37-2, hydrogen nitrate, nital, aqua fortis, azotic acid, acide nitrique, acidum nitricum, rfna, salpetersaeure, nitrous fumes, nitryl hydroxide, 2.5L, CHEBI:48107, Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, HDPE plastic bottle