Check Order Status
Nitric Acid 70% d=1.42, SLR, Extra Pure, Fisher Chemical™
Element Search
Structure Search
Nitric Acid 70% d=1.42, SLR, Extra Pure, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCN2250PB15
also known as FGCN/2250/PB15
1 L
HDPE plastic bottle
Catalog number
FGCN2250PB15
also known as FGCN/2250/PB15
Price (MYR)
293.00
EA
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
293.00
EA
Add to cart
Specifications
pH
1
Color
Yellow
CAS
7697-37-2
Melting Point
-42°C
Boiling Point
122°C
View more