Nitric Acid 65% d=1.40, Certified AR, for Analysis, Fisher Chemical™
Nitric Acid 65% d=1.40, Certified AR, for Analysis, Fisher Chemical™
Nitric Acid 65% d=1.40, Certified AR, for Analysis, Fisher Chemical™
Nitric Acid 65% d=1.40, Certified AR, for Analysis, Fisher Chemical™
Nitric Acid 65% d=1.40, Certified AR, for Analysis, Fisher Chemical™
HNO3, CAS Number-7697-37-2, azotic acid, hydrogen nitrate, aqua fortis, acidum nitricum, acide nitrique, nital, nitrous fumes, salpetersaeure, nitryl hydroxide, rfna, 2.5L, CHEBI:48107, Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, HDPE plastic bottle
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FGCN2185PB17
also known as FGCN/2185/PB17
Price (MYR)
389.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
389.00
EA
Add to cart
Specifications
Chemical Name or MaterialNitric Acid
pH1
ColorYellow
CAS7697-37-2
Melting Point-41°C
View more
HNO3, CAS Number-7697-37-2, azotic acid, hydrogen nitrate, aqua fortis, acidum nitricum, acide nitrique, nital, nitrous fumes, salpetersaeure, nitryl hydroxide, rfna, 2.5L, CHEBI:48107, Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, HDPE plastic bottle