Check Order Status
1-Naphthylamine-7-Sulfonic Acid, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
1-Naphthylamine-7-Sulfonic Acid, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCN091048
also known as FGCN/0910/48
100 g
Plastic powder jar
Catalog number
FGCN091048
also known as FGCN/0910/48
Price (MYR)
-
Quantity:
100 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS
119-28-8
Melting Point
500°C
Color
Beige-Gray
Packaging
Plastic powder jar
Molecular Formula
C10H9NO3S
View more