Check Order Status
Methyl Red, Pure, Solution 0.01%, Indicator Grade, Fisher Chemical™
Element Search
Structure Search
Methyl Red, Pure, Solution 0.01%, Indicator Grade, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCM5171L08
also known as FGCM/5171L/08
500 mL
Catalog number
FGCM5171L08
also known as FGCM/5171L/08
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
MDL Number
2425
Packaging
Amber glass bottle
Quantity
500 mL
Physical Form
Liquid