Check Order Status
Methyl Orange, Pure, Indicator Grade, Solid, C.I.13025, Fisher Chemical™
Element Search
Structure Search
Methyl Orange, Pure, Indicator Grade, Solid, C.I.13025, Fisher Chemical™
Have Questions?
Change view
Quantity:
100 g
25 g
Catalog number
FGCM500048
also known as FGCM/5000/48
Price (MYR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Melting Point
300°C
CAS
547-58-0
Molecular Formula
C14H14N3NaO3S
MDL Number
7502
InChI Key
STZCRXQWRGQSJD-UHFFFAOYSA-M
View more