Check Order Status
Methylene Blue, Extra Pure, SLR, C.I.52015, Fisher Chemical
Element Search
Structure Search
Methylene Blue, Extra Pure, SLR, C.I.52015, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCM485048
also known as FGCM/4850/48
100 g
Catalog number
FGCM485048
also known as FGCM/4850/48
Price (MYR)
446.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
446.00
EA
Add to cart
Specifications
Melting Point
100°C
CAS
7220-79-3
Molecular Formula
C16H24ClN3O3S
MDL Number
MFCD00150008
InChI Key
XQAXGZLFSSPBMK-UHFFFAOYSA-M
View more