Check Order Status
Methyl Benzoate, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Methyl Benzoate, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCM4460PB17
also known as FGCM/4460/PB17
2.5 L
Catalog number
FGCM4460PB17
also known as FGCM/4460/PB17
Price (MYR)
-
Quantity:
2.5 L
Request bulk or custom format
Specifications
CAS
93-58-3
Melting Point
-12°C
Color
Yellow
Boiling Point
198°C
Packaging
Plastic Coated Glass bottle
View more