Check Order Status
Mercury(II) Sulfate, Certified AR for Analysis, Fisher Chemical
Element Search
Structure Search
Mercury(II) Sulfate, Certified AR for Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCM322050
also known as FGCM/3220/50
250 g
Plastic powder jar
Catalog number
FGCM322050
also known as FGCM/3220/50
Price (MYR)
-
Quantity:
250 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS
7783-35-9
Color
White
pH
1
Physical Form
Powder/Solid
Quantity
250 g
View more