Check Order Status
Mercury(II) Oxide, 99+%, Pure, Yellow, Fisher Chemical
Element Search
Structure Search
Mercury(II) Oxide, 99+%, Pure, Yellow, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCM310048
also known as FGCM/3100/48
100 g
Plastic powder jar
Catalog number
FGCM310048
also known as FGCM/3100/48
Price (MYR)
-
Quantity:
100 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
Melting Point
500°C
CAS
21908-53-2
Color
Orange to Yellow
Physical Form
Powder/Solid
Quantity
100 g
View more