Check Order Status
Mercury(II) Acetate, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Mercury(II) Acetate, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCM265050
also known as FGCM/2650/50
Price (MYR)
-
Request bulk or custom format
Specifications
Melting Point
179°C
CAS
1600-27-7
Color
White
pH
4.4
Physical Form
Powder/Solid
View more