Check Order Status
Marble Chips, Pure, Fisher Chemical™
Element Search
Structure Search
Marble Chips, Pure, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCM245063
also known as FGCM/2450/63
3 kg
Catalog number
FGCM245063
also known as FGCM/2450/63
Price (MYR)
-
Quantity:
3 kg
Request bulk or custom format
Specifications
Melting Point
825°C
CAS
471-34-1
Color
White
pH
8 to 9
Physical Form
Solid
View more
Marble Chips, Pure