Check Order Status
Dl-Malic Acid, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Dl-Malic Acid, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCM135053
also known as FGCM/1350/53
500 g
Plastic powder jar
Catalog number
FGCM135053
also known as FGCM/1350/53
Price (MYR)
-
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS
6915-15-7
Melting Point
128°C
Color
White
pH
2
Packaging
Plastic powder jar
View more