Check Order Status
Magnesium Perchlorate, Pure, Dried, Desiccating Agent, Fisher Chemical™
Element Search
Structure Search
Magnesium Perchlorate, Pure, Dried, Desiccating Agent, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCM095050
also known as FGCM/0950/50
250 g
Catalog number
FGCM095050
also known as FGCM/0950/50
Price (MYR)
783.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
783.00
EA
Add to cart
Specifications
Melting Point
250°C
CAS
10034-81-8
Color
White
pH
5 to 9
Physical Form
Solid
View more