Check Order Status
Magnesium Carbonate, Pentahydrated Basic, Extra Pure, Light, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical
Actual product may vary
Magnesium Carbonate, Pentahydrated Basic, Extra Pure, Light, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCM050053
also known as FGCM/0500/53
500 g
Plastic powder jar
Catalog number
FGCM050053
also known as FGCM/0500/53
Price (MYR)
-
Quantity:
500 g
Packaging:
Plastic powder jar
500g, Plastic powder jar
Specifications
CAS
12125-28-9
Color
White
Physical Form
Solid
Quantity
500 g
Molecular Formula
CMgO
3
·1/4H
2
O
2
Mg·5/4H
2
O
MDL Number
149784
Packaging
Plastic powder jar