Check Order Status
Litmus, Pure, Indicator Grade, Fisher Chemical
Element Search
Structure Search
Litmus, Pure, Indicator Grade, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCL250046
also known as FGCL/2500/46
25 g
Catalog number
FGCL250046
also known as FGCL/2500/46
Price (MYR)
737.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
737.00
EA
Add to cart
Specifications
CAS
1393-92-6
MDL Number
131528
Packaging
Glass Jar
Color
Blue
Quantity
25 g
View more