Check Order Status
Linseed Oil, Pure, Raw, ≈d= 0.93
Element Search
Structure Search
Linseed Oil, Pure, Raw, ≈d= 0.93
Have Questions?
Catalog Number
Quantity
FGCL195017
also known as FGCL/1950/17
2.5 L
Catalog number
FGCL195017
also known as FGCL/1950/17
Price (MYR)
-
Quantity:
2.5 L
Request bulk or custom format
Specifications
CAS
8001-26-1
Melting Point
-23.9°C
Color
Yellow
Boiling Point
315.6°C
Packaging
Amber glass bottle
View more