Check Order Status
Karl Fischer Aqualine™ Sodium Tartrate-2H2O, for Karl Fischer Titration, Fisher Chemical
Element Search
Structure Search
Karl Fischer Aqualine™ Sodium Tartrate-2H2O, for Karl Fischer Titration, Fisher Chemical
Solid standard
Have Questions?
Catalog Number
Quantity
FGCK277048
also known as FGCK/2770/48
100 g
Catalog number
FGCK277048
also known as FGCK/2770/48
Price (MYR)
530.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
530.00
BOT
Add to cart
Specifications
Chemical Name or Material
Aqualine™ Sodium Tartrate Dihydrate Water Standard
Color
White
Quantity
100 g
Name Note
Karl Fischer Reagent
CAS
6106-24-7
View more
Primary solid water standard for Karl Fischer volumetric analysis
Contains 15.66% ± 0.05% water