Check Order Status
Karl Fischer Aqualine™ Water Standard 5mg, for Karl Fischer Titration, Fisher Chemical
Element Search
Structure Search
Karl Fischer Aqualine™ Water Standard 5mg, for Karl Fischer Titration, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCK273008
also known as FGCK/2730/08
500 mL
Catalog number
FGCK273008
also known as FGCK/2730/08
Price (MYR)
528.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
528.00
EA
Add to cart
Specifications
Quantity
500 mL
Packaging
Amber glass bottle
Physical Form
Liquid
MDL Number
240423