Karl Fischer Aqualine™ Electrolyte CG, for Karl Fischer Titration By Coulometry, Fisher Chemical
Karl Fischer Aqualine™ Electrolyte CG, for Karl Fischer Titration By Coulometry, Fisher Chemical
Actual product may vary
Karl Fischer Aqualine™ Electrolyte CG, for Karl Fischer Titration By Coulometry, Fisher Chemical
Have Questions?
Catalog NumberQuantity
FGCK256004
also known as FGCK/2560/04
25 mL
Catalog number FGCK256004
also known as FGCK/2560/04
Price (MYR)
270.00
EA
Add to cart
Quantity:
25 mL
Price (MYR)
270.00
EA
Add to cart
25mL, Vial
  • Catholyte solution for use in conventional cells with a diaphragm
  • Reagent required to complete the electrochemical reaction in the cathode compartment for coulometric analysis
  • Contains methanol as a solvent
Specifications
Chemical Name or MaterialAqualine™ Electrolyte CG
ColorColorless
Quantity25 mL
Name NoteKarl Fischer Reagent
ChemAlert Storage SymbolRed
Health Hazard 2DANGER!
CAS108-75-8
Health Hazard 3Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .
NFPA
Health:1
Flammability:3
Instability:0
Boiling Point64°C
PackagingVial
CAS7446-09-5
Physical FormLiquid
CAS7553-56-2
Use TestPass Test
MDL Number3097388
Unit SizeEA