Check Order Status
Karl Fischer Aqualine™ Electrolyte AD-G, for Karl Fischer Titration By Coulometry, Fisher Chemical
Element Search
Structure Search
Karl Fischer Aqualine™ Electrolyte AD-G, for Karl Fischer Titration By Coulometry, Fisher Chemical
Electrolyte solution for use in cells without a diaphragm
Have Questions?
Catalog Number
Quantity
FGCK251508
also known as FGCK/2515/08
500 mL
Catalog number
FGCK251508
also known as FGCK/2515/08
Price (MYR)
499.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
499.00
BOT
Add to cart
Chemical Identifiers
CAS
108-75-8, 7446-09-5, 124-68-5
Specifications
Chemical Name or Material
Aqualine™ Electrolyte ADG
Quantity
500 mL
Cleanup Symbol
Solvent
Name Note
Karl Fisher Reagent
ChemAlert Storage Symbol
Red
View more