Check Order Status
Karl Fischer Aqualine™ Electrolyte AD, for Karl Fischer Titration By Coulometry, Fisher Chemical
Actual product may vary
Karl Fischer Aqualine™ Electrolyte AD, for Karl Fischer Titration By Coulometry, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCK251008
also known as FGCK/2510/08
500 mL
Catalog number
FGCK251008
also known as FGCK/2510/08
Price (MYR)
489.00
EA
Add to cart
Quantity:
500 mL
Price (MYR)
489.00
EA
Add to cart
500mL, Amber glass bottle
Specifications
Quantity
500 mL
Packaging
Amber glass bottle
Physical Form
Liquid
MDL Number
81734