Check Order Status
Karl Fischer Aqualine™ Titrant 5, for Karl Fischer Titration By Volumetry, Fisher Chemical
Element Search
Structure Search
Karl Fischer Aqualine™ Titrant 5, for Karl Fischer Titration By Volumetry, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCK220017
also known as FGCK/2200/17
2.5 L
Catalog number
FGCK220017
also known as FGCK/2200/17
Price (MYR)
884.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
884.00
EA
Add to cart
Specifications
Chemical Name or Material
Aqualine™ Titrant 5
Color
Colorless
Purity or Free of Statement
Pyridine free
Quantity
2.5 L
Cleanup Symbol
Solvent
View more