Check Order Status
Kieselguhr, Pure, Washed with Acid, Fisher Chemical™
Element Search
Structure Search
Kieselguhr, Pure, Washed with Acid, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 kg
500 g
Packaging:
View all
Plastic Keg
Catalog number
FGCK010060
also known as FGCK/0100/60
Price (MYR)
1,613.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic Keg
Request bulk or custom format
Price (MYR)
1,613.00
EA
Add to cart
Chemical Identifiers
CAS
61790-53-2
Specifications
Physical Form
Solid
Color
Gray
Quantity
1 kg
pH
9 to 10
CAS
61790-53-2
View more