Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution, Ready to Use, for Volumetric Analysis, meets analytical spec of Ph.Eur., BP, Fisher Chemical™
Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution, Ready to Use, for Volumetric Analysis, meets analytical spec of Ph.Eur., BP, Fisher Chemical™
Actual product may vary
Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution, Ready to Use, for Volumetric Analysis, meets analytical spec of Ph.Eur., BP, Fisher Chemical™
Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution, Ready to Use, for Volumetric Analysis, meets analytical spec of Ph.Eur., BP, Fisher Chemical™
Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution, Ready to Use, for Volumetric Analysis, meets analytical spec of Ph.Eur., BP, Fisher Chemical™
Have Questions?
Catalog NumberQuantityPackaging
FGCJ843015
also known as FGCJ/8430/15
1 LHDPE plastic bottle
Catalog number FGCJ843015
also known as FGCJ/8430/15
Price (MYR)
-
Quantity:
1 L
Packaging:
HDPE plastic bottle
Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP
Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP
Specifications
pH1.5
ColorColorless
CAS7664-93-9
Boiling Point100°C
PackagingHDPE plastic bottle
Vapor Pressure1mmHg at 20°C
Quantity1 L
Molecular FormulaH2O4S
Physical FormLiquid
MDL NumberMFCD00064589
Synonymoil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
IUPAC Namesulfuric acid
Molecular Weight (g/mol)98.07
PubChem CID1118
ChEBICHEBI:26836